accessibility menu, dialog, popup

UserName

NaB

CAS RN 143-66-8

IUPAC Name: sodium tetraphenylboranuide
Synonyms: sodium tetraphenylborate
Molecular Weight (g/mol): 342.22
Molecular Formula: C24H20BNa
InChi Key: HFSRCEJMTLMDLI-UHFFFAOYSA-N
SMILES: [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (7)
Grade 
  • (3)
  • (8)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (11)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (7)
  • (3)
  • (8)
  • (1)
  • (1)