missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraphenylborate, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18613895
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613895 5 kg Amber Glass Bottle
18613891 10 g Amber Glass Bottle
18613892 25 g Amber Glass Bottle
18613893 100 g Amber Glass Bottle
18613894 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613895 Supplier Spectrum Chemical Mfg Cor Supplier No. S14905KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143-66-8

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 5 kg
Molecular Weight (g/mol) 342.22
Percent Purity 99.5%
Grade ACS/Reagent
Loss on Drying 0.005
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Tetraphenylborate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.