accessibility menu, dialog, popup

UserName

CAS RN 14267-92-6

IUPAC Name: 5-chloropent-1-yne
Synonyms: 5-chloro-1-pentyne
Molecular Weight (g/mol): 102.56
Molecular Formula: C5H7Cl
InChi Key: UXFIKVWAAMKFQE-UHFFFAOYSA-N
SMILES: ClCCCC#C
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)