missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-pentyne 96.0+%, TCI America™

Catalog No. C15225ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15225ML 5 mL
C152225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15225ML Supplier TCI America Supplier No. C15225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14267-92-6
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.561
MDL Number MFCD00001014
InChI Key UXFIKVWAAMKFQE-UHFFFAOYSA-N
Synonym 5-chloro-1-pentyne, 1-pentyne, 5-chloro, 5-chloropentyne, 4-pentynyl chloride, 1-chloro-4-pentine, 1-chloro-4-pentyne, pent-4-ynyl chloride, 5-chloro-pent-1-yne, acmc-1bzfd, pent-4-yn-1-yl chloride
PubChem CID 84308
IUPAC Name 5-chloropent-1-yne
SMILES C#CCCCCl

Specifications

Color Yellow
Boiling Point 113°C
Quantity 5 mL
UN Number 1993
Formula Weight 102.56
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-1-pentyne