accessibility menu, dialog, popup

UserName

CAS RN 142072-12-6

IUPAC Name: N-[2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Synonyms: N-[2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular Weight (g/mol): 290.28
Molecular Formula: C10H18N4O6
InChi Key: DHQJPCIPZHPDSL-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(O)C(CO)OC1OCCN=[N+]=[N-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results