missing translation for 'onlineSavingsMsg'
Learn More

2-Azidoethyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. A2172500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2172500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2172500MG Supplier TCI America Supplier No. A2172500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142072-12-6
Molecular Formula C10H18N4O6
Molecular Weight (g/mol) 290.276
InChI Key DHQJPCIPZHPDSL-SRQGCSHVSA-N
PubChem CID 44630006
IUPAC Name N-[(2R,3S,4R,5S,6S)-2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1OCCN=[N+]=[N-])CO)O)O

Specifications

Color White
Quantity 500 mg
Formula Weight 290.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Azidoethyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer