accessibility menu, dialog, popup

UserName

OOO

CAS RN 14166-21-3

IUPAC Name: octahydro-2-benzofuran-1,3-dione
Synonyms: hexahydrophthalic anhydride HHPA
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: MUTGBJKUEZFXGO-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2CCCCC12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results