CAS RN 141412-60-4
IUPAC Name:
1,4-difluoro-2-methyl-5-nitrobenzene
Synonyms:
1,4-difluoro-2-methyl-5-nitrobenzene
Molecular Weight (g/mol):
173.12
Molecular Formula:
C7H5F2NO2
InChi Key:
ABWWQMGGJXKGBL-UHFFFAOYSA-N
SMILES:
CC1=CC(F)=C(C=C1F)[N+]([O-])=O
1
–
1
of
1
results
1
2,5-Difluoro-4-nitrotoluene 98.0+%, TCI America™
CAS: 141412-60-4 Molecular Formula: C7H5F2NO2 Molecular Weight (g/mol): 173.12 MDL Number: MFCD06200905 InChI Key: ABWWQMGGJXKGBL-UHFFFAOYSA-N Synonym: 2,5-difluoro-4-nitrotoluene,2,5-difluoro-4-methylnitrobenzene,2,5-difluoro-4-methyl nitrobenzene,benzene, 1,4-difluoro-2-methyl-5-nitro,2,5-difluoro-1-methyl-4-nitrobenzene,1,4-difluoro-2-methyl-5-nitro-benzene,pubchem2555,difluoromethylnitrobenzene,ksc495e0j,acmc-1c025 PubChem CID: 230658 IUPAC Name: 1,4-difluoro-2-methyl-5-nitrobenzene SMILES: CC1=CC(F)=C(C=C1F)[N+]([O-])=O