missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-nitrotoluene 98.0+%, TCI America™

Catalog No. D48065G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48065G 5 g
D48061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48065G Supplier TCI America Supplier No. D48065G

Chemical Identifiers

CAS 141412-60-4
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.12
MDL Number MFCD06200905
InChI Key ABWWQMGGJXKGBL-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-nitrotoluene, 2,5-difluoro-4-methylnitrobenzene, 2,5-difluoro-4-methyl nitrobenzene, benzene, 1,4-difluoro-2-methyl-5-nitro, 2,5-difluoro-1-methyl-4-nitrobenzene, 1,4-difluoro-2-methyl-5-nitro-benzene, pubchem2555, difluoromethylnitrobenzene, ksc495e0j, acmc-1c025
PubChem CID 230658
IUPAC Name 1,4-difluoro-2-methyl-5-nitrobenzene
SMILES CC1=CC(F)=C(C=C1F)[N+]([O-])=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 173.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Difluoro-4-nitrotoluene