accessibility menu, dialog, popup

UserName

CAS RN 14080-23-0

IUPAC Name: pyrimidine-2-carbonitrile
Synonyms: 2-pyrimidinecarbonitrile
Molecular Weight (g/mol): 105.1
Molecular Formula: C5H3N3
InChi Key: IIHQNAXFIODVDU-UHFFFAOYSA-N
SMILES: N#CC1=NC=CC=N1
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)