accessibility menu, dialog, popup

UserName

KFBFFF

CAS RN 14075-53-7

IUPAC Name: potassium tetrafluoroboranuide
Synonyms: potassium tetrafluoroborate
Molecular Weight (g/mol): 125.9
Molecular Formula: BF4K
InChi Key: AKEBROIVCDHVSD-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)F
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (8)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)