missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrafluoroborate, 99%, extra pure

Catalog No. AC269220250
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269220250 25 g Plastic bottle
AC269220010 1 kg Plastic bottle
AC269220050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC269220250 Supplier Thermo Scientific Chemicals Supplier No. 269220250

CAS: 14075-53-7 | BF4K | 125.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 14075-53-7
Molecular Formula BF4K
Molecular Weight (g/mol) 125.90
MDL Number MFCD00011395
InChI Key AKEBROIVCDHVSD-UHFFFAOYSA-N
Synonym potassium tetrafluoroborate, potassium fluoroborate, potassium fluoborate, potassium borofluoride, avogadrite, potassium boron fluoride, potassium fluorohydroborate, potassium hydrofluoroborate, boron potassium fluoride, potassium boride fluoride
PubChem CID 518872
IUPAC Name potassium tetrafluoroboranuide
SMILES [K+].F[B-](F)(F)F

Specifications

Melting Point 530.0°C
Color White to Gray
Physical Form Micro-crystalline Powder
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula KBF4
Merck Index 15,7804
Solubility Information Solubility in water: 4.4g/L (20°C). Other solubilities: slightly soluble in boiling alcohol
Formula Weight 125.91
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Density 2.5
Chemical Name or Material Potassium tetrafluoroborate, c.p.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-928-2
RTECSNumber ED2800000
TSCA TSCA

RUO – Research Use Only