accessibility menu, dialog, popup

UserName

CAS RN 140-53-4

IUPAC Name: 2-(4-chlorophenyl)acetonitrile
Synonyms: 4-chlorobenzyl cyanide C8H6ClN
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: IVYMIRMKXZAHRV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CC#N)C=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)