missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl Cyanide 98.0+%, TCI America™

Catalog No. C0585500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0585500G 500 g
C058525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0585500G Supplier TCI America Supplier No. C0585500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-53-4
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001918
InChI Key IVYMIRMKXZAHRV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
PubChem CID 241582
ChEBI CHEBI:17346
IUPAC Name 2-(4-chlorophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)Cl

Specifications

Color White-Yellow
Boiling Point 267°C
Quantity 500 g
UN Number 3439
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chlorobenzyl Cyanide
EINECSNumber (3)-1784
RTECSNumber AL8250000
TSCA Yes