accessibility menu, dialog, popup

UserName

CAS RN 140-08-9

ClOPOClOCl

CAS RN 140-08-9

IUPAC Name: tris(2-chloroethyl) phosphite
Synonyms: tris(2-chloroethyl) phosphite
Molecular Weight (g/mol): 269.48
Molecular Formula: C6H12Cl3O3P
InChi Key: LUVCTYHBTXSAMX-UHFFFAOYSA-N
SMILES: ClCCOP(OCCCl)OCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results