missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-chloroethyl) Phosphite 93.0+%, TCI America™

Catalog No. P027725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P027725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P027725ML Supplier TCI America Supplier No. P027725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-08-9
Molecular Formula C6H12Cl3O3P
Molecular Weight (g/mol) 269.483
MDL Number MFCD00000968
InChI Key LUVCTYHBTXSAMX-UHFFFAOYSA-N
Synonym Phosphorous Acid Tris(2-chloroethyl) Ester
PubChem CID 8783
IUPAC Name tris(2-chloroethyl) phosphite
SMILES C(CCl)OP(OCCCl)OCCCl

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 mL
UN Number 2810
Formula Weight 269.48
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-chloroethyl) Phosphite
EINECSNumber (9)-0893
RTECSNumber KK2810000
TSCA Yes