accessibility menu, dialog, popup

UserName

CAS RN 13978-85-3

IUPAC Name: zinc(2+) bis(quinolin-8-olate)
Synonyms: zinc(2+) bis(quinolin-8-olate)
Molecular Weight (g/mol): 353.69
Molecular Formula: C18H12N2O2Zn
InChi Key: HTPBWAPZAJWXKY-UHFFFAOYSA-L
SMILES: [Zn++].[O-]C1=C2N=CC=CC2=CC=C1.[O-]C1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results