missing translation for 'onlineSavingsMsg'
Learn More

Bis(8-quinolinolato)zinc(II) Hydrate 93.0+%, TCI America™

Catalog No. B16785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16785G 5 g
B167825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16785G Supplier TCI America Supplier No. B16785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13978-85-3
Molecular Formula C18H12N2O2Zn
Molecular Weight (g/mol) 353.686
MDL Number MFCD00065291
InChI Key HTPBWAPZAJWXKY-UHFFFAOYSA-L
Synonym zinc 8-quinolinolate, zinc oxyquinolate, bis 8-quinolinolato zinc, zinc quinolin-8-olate, unii-2p1rtx6vak, bis 8-hydroxyquinolinato zinc, 2p1rtx6vak, bis 8-quinolinolato zinc ii, bis 8-hydroxyquinolinato zinc ii, 8-hydroxyquinoline, zinc salt
PubChem CID 84127
IUPAC Name zinc;quinolin-8-olate
SMILES C1=CC2=C(C(=C1)[O-])N=CC=C2.C1=CC2=C(C(=C1)[O-])N=CC=C2.[Zn+2]

Specifications

Color Green-Yellow
Quantity 5 g
Formula Weight 353.69
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(8-quinolinolato)zinc(II) Hydrate
EINECSNumber (5)-0806
TSCA Yes