CAS RN 13965-03-2
trans-Dichlorobis(triphenylphosphine)palladium(II), Premion™, 99.95% (metals basis), Pd 14.7% min
CAS: 13965-03-2 Molecular Formula: C36H30Cl2P2Pd Molecular Weight (g/mol): 701.90 MDL Number: MFCD00009593 InChI Key: YNHIGQDRGKUECZ-UHFFFAOYSA-L Synonym: bis triphenylphosphine palladium ii dichloride,bis triphenylphosphine palladium ii chloride,dichlorobis triphenylphosphine palladium,bis triphenylphosphine palladium chloride,bis triphenylphosphine palladiuml ii dichloride,palladium 2+ ; triphenylphosphane; dichloride,trans-dichlorobis triphenylphosphine palladium ii 1g PubChem CID: 131664180 IUPAC Name: ethane;methane;palladium(2+);triphenylphosphane;dichloride SMILES: [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
CAS: 13965-03-2 Molecular Formula: C36H30Cl2P2Pd Molecular Weight (g/mol): 701.90 MDL Number: MFCD00009593 InChI Key: YNHIGQDRGKUECZ-UHFFFAOYSA-L Synonym: bis triphenylphosphine palladium ii dichloride,bis triphenylphosphine palladium ii chloride,dichlorobis triphenylphosphine palladium,bis triphenylphosphine palladium chloride,bis triphenylphosphine palladiuml ii dichloride,palladium 2+ ; triphenylphosphane; dichloride,trans-dichlorobis triphenylphosphine palladium ii 1g PubChem CID: 131664180 IUPAC Name: ethane;methane;palladium(2+);triphenylphosphane;dichloride SMILES: [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd
CAS: 13965-03-2 Molecular Formula: C36H30Cl2P2Pd Molecular Weight (g/mol): 701.90 MDL Number: MFCD00009593 InChI Key: YNHIGQDRGKUECZ-UHFFFAOYSA-L Synonym: bis triphenylphosphine palladium ii dichloride,bis triphenylphosphine palladium ii chloride,dichlorobis triphenylphosphine palladium,bis triphenylphosphine palladium chloride,bis triphenylphosphine palladiuml ii dichloride,palladium 2+ ; triphenylphosphane; dichloride,trans-dichlorobis triphenylphosphine palladium ii 1g PubChem CID: 131664180 IUPAC Name: ethane;methane;palladium(2+);triphenylphosphane;dichloride SMILES: [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Bis(triphenylphosphine)palladium(II) chloride, 98%
CAS: 13965-03-2 Molecular Formula: C36H30Cl2P2Pd Molecular Weight (g/mol): 701.90 MDL Number: MFCD00009593 InChI Key: YNHIGQDRGKUECZ-UHFFFAOYSA-L Synonym: bis triphenylphosphine palladium ii dichloride,bis triphenylphosphine palladium ii chloride,dichlorobis triphenylphosphine palladium,bis triphenylphosphine palladium chloride,bis triphenylphosphine palladiuml ii dichloride,palladium 2+ ; triphenylphosphane; dichloride,trans-dichlorobis triphenylphosphine palladium ii 1g PubChem CID: 131664180 IUPAC Name: ethane;methane;palladium(2+);triphenylphosphane;dichloride SMILES: [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Bis(triphenylphosphine)palladium(II) Dichloride 98.0+%, TCI America™
CAS: 13965-03-2 Molecular Formula: C36H30Cl2P2Pd Molecular Weight (g/mol): 701.90 MDL Number: MFCD00009593 InChI Key: YNHIGQDRGKUECZ-UHFFFAOYSA-L Synonym: bis triphenylphosphine palladium ii dichloride,bis triphenylphosphine palladium ii chloride,dichlorobis triphenylphosphine palladium,bis triphenylphosphine palladium chloride,bis triphenylphosphine palladiuml ii dichloride,palladium 2+ ; triphenylphosphane; dichloride,trans-dichlorobis triphenylphosphine palladium ii 1g PubChem CID: 131664180 IUPAC Name: palladium(2+) bis(triphenylphosphane) dichloride SMILES: [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1