accessibility menu, dialog, popup

UserName

CAS RN 1391728-13-4

IUPAC Name: 1-fluoro-3,3-dimethyl-3H-1λ³,2-benziodaoxole
Synonyms: 1-fluoro-3,3-dimethyl-1λ³,2-benziodaoxole
Molecular Weight (g/mol): 280.08
Molecular Formula: C9H10FIO
InChi Key: FISITPXJYDTLHI-UHFFFAOYSA-N
SMILES: CC1(C)O[I](F)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results