accessibility menu, dialog, popup

UserName

CAS RN 139-07-1

IUPAC Name: benzyl(dodecyl)dimethylazanium chloride
Synonyms: benzododecinium chloride 苯度氯铵
Molecular Weight (g/mol): 339.99
Molecular Formula: C21H38ClN
InChi Key: JBIROUFYLSSYDX-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)