missing translation for 'onlineSavingsMsg'
Learn More

Benzyldodecyldimethylammonium Chloride Dihydrate 98.0+%, TCI America™

Catalog No. B217125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B217125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B217125G Supplier TCI America Supplier No. B217125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139-07-1
Molecular Formula C21H38ClN
Molecular Weight (g/mol) 339.99
MDL Number MFCD00137276
InChI Key JBIROUFYLSSYDX-UHFFFAOYSA-M
Synonym benzyldimethyldodecylammonium chloride, benzododecinium chloride, n-benzyl-n,n-dimethyldodecan-1-aminium chloride, cequartyl a, catigene om, benzyldodecyldimethylammonium chloride, zephirol, dehyquart ldb, catiogen pan, vantoc cl
PubChem CID 8753
IUPAC Name benzyl(dodecyl)dimethylazanium chloride
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point 44°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 339.99
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyldodecyldimethylammonium Chloride Dihydrate
EINECSNumber (3)-2694
RTECSNumber BO6821500
TSCA Yes