accessibility menu, dialog, popup

UserName

CAS RN 13831-03-3

IUPAC Name: tert-butyl prop-2-ynoate
Synonyms: tert-butyl prop-2-ynoate
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: XGTPDIIFEPTULX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C#C
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)