missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl propiolate, 98%

Catalog No. AC435390010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435390010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435390010 Supplier Thermo Scientific Chemicals Supplier No. 435390010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13831-03-3 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13831-03-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00060100
InChI Key XGTPDIIFEPTULX-UHFFFAOYSA-N
Synonym tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
PubChem CID 543038
IUPAC Name tert-butyl prop-2-ynoate
SMILES CC(C)(C)OC(=O)C#C

Specifications

Melting Point 18°C to 20°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 52°C to 53°C (27.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.92
Formula Weight 126.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butyl propiolate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only