accessibility menu, dialog, popup

UserName

H3CONOHHNOONOHNHOONOHNH2CH3SOHOO

CAS RN 138-14-7

IUPAC Name: N-(5-aminopentyl)-N-hydroxy-N'-[5-(N-hydroxy-3-{[5-(N-hydroxyacetamido)pentyl]carbamoyl}propanamido)pentyl]butanediamide; methanesulfonic acid
Synonyms: deferoxamine; methanesulfonic acid
Molecular Weight (g/mol): 656.79
Molecular Formula: C26H52N6O11S
InChi Key: IDDIJAWJANBQLJ-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCN
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)