missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Deferoxamine mesylate, 95%

Catalog No. AC461770010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461770010 Supplier Thermo Scientific Chemicals Supplier No. 461770010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 138-14-7 | C25H48N6O8·CH4O3S | 656.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138-14-7
Molecular Formula C25H48N6O8·CH4O3S
Molecular Weight (g/mol) 656.79
InChI Key IDDIJAWJANBQLJ-UHFFFAOYSA-N
PubChem CID 62881
ChEBI CHEBI:31460

Specifications

Melting Point 148°C to 149°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Colorless to light brown-yellow
Formula Weight 656.79
Percent Purity 95%
Chemical Name or Material Deferoxamine mesylate
EINECSNumber 205-314-3

RUO – Research Use Only