accessibility menu, dialog, popup

UserName

CAS RN 137407-31-9

IUPAC Name: 4-methoxyphenyl 4-[(6-hydroxyhexyl)oxy]benzoate
Synonyms: 4-methoxyphenyl 4-[(6-hydroxyhexyl)oxy]benzoate
Molecular Weight (g/mol): 344.41
Molecular Formula: C20H24O5
InChi Key: KTTALHXKXPIECT-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(=O)C2=CC=C(OCCCCCCO)C=C2)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results