missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 4-(6-Hydroxyhexyloxy)benzoate, TCI America™

Catalog No. H0766100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0766100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0766100MG Supplier TCI America Supplier No. H0766100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137407-31-9
Molecular Formula C20H24O5
Molecular Weight (g/mol) 344.41
MDL Number MFCD00191513
InChI Key KTTALHXKXPIECT-UHFFFAOYSA-N
Synonym 4-(6-Hydroxyhexyloxy)benzoic Acid 4-Methoxyphenyl Ester
PubChem CID 44630339
IUPAC Name 4-methoxyphenyl 4-[(6-hydroxyhexyl)oxy]benzoate
SMILES COC1=CC=C(OC(=O)C2=CC=C(OCCCCCCO)C=C2)C=C1

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 100 mg
Formula Weight 344.41
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 4-(6-Hydroxyhexyloxy)benzoate