accessibility menu, dialog, popup

UserName

CAS RN 137156-41-3

IUPAC Name: ethyl 1-hydroxy-1H-1,2,3-triazole-4-carboxylate
Synonyms: ethyl 1-hydroxy-1,2,3-triazole-4-carboxylate
Molecular Weight (g/mol): 157.13
Molecular Formula: C5H7N3O3
InChi Key: FIRHQRGFVOSDDY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN(O)N=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results