missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-Hydroxy-1H-1,2,3-triazole-4-carboxylate 98.0+%, TCI America™

Catalog No. E09015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09015G 5 g
E09011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09015G Supplier TCI America Supplier No. E09015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137156-41-3
Molecular Formula C5H7N3O3
Molecular Weight (g/mol) 157.129
MDL Number MFCD06796746
InChI Key FIRHQRGFVOSDDY-UHFFFAOYSA-N
Synonym 1-Hydroxy-1H-1,2,3-triazole-4-carboxylic Acid Ethyl Ester
PubChem CID 5270538
IUPAC Name ethyl 1-hydroxytriazole-4-carboxylate
SMILES CCOC(=O)C1=CN(N=N1)O

Specifications

Melting Point 105°C
Color White
Quantity 5 g
Formula Weight 157.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-Hydroxy-1H-1,2,3-triazole-4-carboxylate