accessibility menu, dialog, popup

UserName

HClH3CHNOONCH3CH3

CAS RN 136-47-0

IUPAC Name: hydrogen 2-(dimethylamino)ethyl 4-(butylamino)benzoate chloride
Synonyms: tetracaine hydrochloride
Molecular Weight (g/mol): 300.83
Molecular Formula: C15H25ClN2O2
InChi Key: PPWHTZKZQNXVAE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results