accessibility menu, dialog, popup

UserName

CAS RN 13598-65-7

NH4OReOOO

CAS RN 13598-65-7

IUPAC Name: rheniumoylol amine
Synonyms: perrhenic acid amine
Molecular Weight (g/mol): 268.24
Molecular Formula: H4NO4Re
InChi Key: XPSXHXWHJAIQJR-UHFFFAOYSA-M
SMILES: N.O[Re](=O)(=O)=O
Grade 
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)