missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Rhenium Standard for ICP 10 g/L in H2O, Ricca Chemical
Description
- 10 g/L in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS | 13598-65-7, 7732-18-5 |
| CAS Min % | 96.59%, 1.41% |
| CAS Max % | 100%, 1.47% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| Molecular Formula | H4NO4Re |
| MDL Number | MFCD00012377 |
| Color | Colorless |
| Melting Point | Approximately 0°C |
| Show More |