missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Rhenium Standard for ICP 1000 ppm in H2O, Ricca Chemical
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 97.86%, 0.14% |
| CAS Max % | 100%, 0.14% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 500 mL |
| Grade | High Purity |