missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium ICP Standard, 1000 ppm Re in H2O, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 1,000ppm Re |
| CAS Min % | 98.0, 0.14 |
| CAS Max % | 100.0, 0.15 |
| Equivalence | 1 mL = 1 mg Re |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Type | Standard |
| Quantity | 500 mL |
| Show More |