missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Beryllium Standard for ICP 10 g/L in 5% HNO3/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010439100N
Description
- 10 g/L in 5% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Beryllium ICP Standard | |
| 13597-99-4 , 7697-37-2 , 7664-39-3 , 7732-18-5 | |
| 80.82%,15.06%,5.10%,1.02% | |
| Liquid | |
| UN2922 | |
| <2 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| RFVVBBUVWAIIBT-UHFFFAOYSA-N | |
| beryllium;dinitrate | |
| 100 mL | |
| High Purity |
| Natural LDPE Bottle | |
| 77.66%,14.46%,4.90%,0.98% | |
| 1 | |
| BeN2O6 | |
| Colorless | |
| Approximately 0°C | |
| Miscible with water | |
| Standard | |
| [Be+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-] | |
| 133.02 | |
| 26126 |