CAS RN 135841-49-5
CAS RN 135841-49-5
IUPAC Name:
trimethyl(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)silane
Synonyms:
trimethyl(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)silane
Molecular Weight (g/mol):
392.24
Molecular Formula:
C9H9F13Si
InChi Key:
AHGSEBBCGYXEBF-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
1
of
1
results
1
Trimethyl(tridecafluorohexyl)silane 97.0+%, TCI America™
CAS: 135841-49-5 Molecular Formula: C9H9F13Si Molecular Weight (g/mol): 392.24 MDL Number: MFCD20489406 InChI Key: AHGSEBBCGYXEBF-UHFFFAOYSA-N Synonym: Trimethyl(perfluorohexyl)silane PubChem CID: 10894469 IUPAC Name: trimethyl(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)silane SMILES: C[Si](C)(C)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F