accessibility menu, dialog, popup

UserName

H3COOSCH3

CAS RN 13532-18-8

IUPAC Name: methyl 3-(methylsulfanyl)propanoate
Synonyms: methyl 3-(methylsulfanyl)propanoate
Molecular Weight (g/mol): 134.19
Molecular Formula: C5H10O2S
InChi Key: DMMJVMYCBULSIS-UHFFFAOYSA-N
SMILES: COC(=O)CCSC
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)