missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(methylthio)propionate, 98%

Catalog No. AAA1064936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1064936 500 g
AAA1064914 25 g
AAA1064922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1064936 Supplier Thermo Scientific Chemicals Supplier No. A1064936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13532-18-8 | C5H10O2S | 134.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13532-18-8
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00008565
InChI Key DMMJVMYCBULSIS-UHFFFAOYSA-N
Synonym methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate
PubChem CID 61641
IUPAC Name methyl 3-methylsulfanylpropanoate
SMILES COC(=O)CCSC

Specifications

Density 1.074
Boiling Point 72°C to 73°C (11 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.465
Linear Formula CH3SCH2CH2CO2CH3
Quantity 500 g
Beilstein 1745077
Formula Weight 134.2
Percent Purity 98%
Chemical Name or Material Methyl 3- (methylthio)propionate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 236-883-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only