accessibility menu, dialog, popup

UserName

CAS RN 13446-09-8

NH4OIOO

CAS RN 13446-09-8

IUPAC Name: ammonium iodate
Synonyms: ammonium iodate
Molecular Weight (g/mol): 192.94
Molecular Formula: H4INO3
InChi Key: ZRDJERPXCFOFCP-UHFFFAOYSA-N
SMILES: [NH4+].[O-][I](=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results