accessibility menu, dialog, popup

UserName

BrBr

CAS RN 13440-24-9

IUPAC Name: (1,2-dibromo-2-phenylethyl)benzene
Synonyms: (1,2-dibromo-2-phenylethyl)benzene
Molecular Weight (g/mol): 340.06
Molecular Formula: C14H12Br2
InChi Key: GKESIQQTGWVOLH-UHFFFAOYNA-N
SMILES: BrC(C(Br)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)