accessibility menu, dialog, popup

UserName

CH3OH3COH3COH3COHf

CAS RN 13428-80-3

IUPAC Name: hafnium(4+) tetraethanolate
Synonyms: hafnium(4+) tetrakis(ethoxide)
Molecular Weight (g/mol): 358.73
Molecular Formula: C8H20HfO4
InChi Key: BFIMXCBKRLYJQO-UHFFFAOYSA-N
SMILES: [Hf+4].CC[O-].CC[O-].CC[O-].CC[O-]
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results