missing translation for 'onlineSavingsMsg'
Learn More

Hafnium ethoxide, 99.9% (metals basis)

Catalog No. AA4160714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4160714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4160714 Supplier Thermo Scientific Chemicals Supplier No. 04160714

Preparation of refractory coatings | CAS: 13428-80-3 | C8H24HfO4 | 362.766 g/mol

Hafnium ethoxide is a chemical used in the preparation of refractory coatings. Some of the other applications include Nano-materials & Thin Films, Display & Imaging, Catalysis & Synthesis, Hybrid & Disperse Materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13428-80-3
Molecular Formula C8H24HfO4
Molecular Weight (g/mol) 362.766
MDL Number MFCD00070459
InChI Key ZSVBAHMGGMMOSD-UHFFFAOYSA-N
Synonym hafnium tetraethoxide, hafnium iv ethoxide
PubChem CID 72429791
IUPAC Name ethanol;hafnium
SMILES CCO.CCO.CCO.CCO.[Hf]

Specifications

Melting Point 178°C to 180°C
Boiling Point 180°C to 200°C (13 mmHg)
Quantity 25 g
UN Number UN3180
Sensitivity Moisture sensitive
Solubility Information It is soluble in hydrocarbons,reacts with ketones,esters. Reacts exothermally with water
Formula Weight 358.73
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Hafnium ethoxide
Show More Show Less
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only