accessibility menu, dialog, popup

UserName

CAS RN 1338446-77-7

NO

CAS RN 1338446-77-7

IUPAC Name: 9-(3-{8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl}phenyl)-9H-carbazole
Synonyms: 9-(3-{8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl}phenyl)carbazole
Molecular Weight (g/mol): 409.49
Molecular Formula: C30H19NO
InChi Key: YEWVLWWLYHXZLZ-UHFFFAOYSA-N
SMILES: O1C2=C(C=CC=C2)C2=C1C=CC(=C2)C1=CC(=CC=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results