missing translation for 'onlineSavingsMsg'
Learn More

9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole 98.0+%, TCI America™

Catalog No. D4919200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4919200MG 200 mg
D49191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4919200MG Supplier TCI America Supplier No. D4919200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1338446-77-7
Molecular Formula C30H19NO
Molecular Weight (g/mol) 409.488
InChI Key YEWVLWWLYHXZLZ-UHFFFAOYSA-N
PubChem CID 58556666
IUPAC Name 9-(3-dibenzofuran-2-ylphenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=CC(=C4)C5=CC6=C(C=C5)OC7=CC=CC=C76

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 409.49
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole