accessibility menu, dialog, popup

UserName

H3CCH3NNNCH3OClOOO

CAS RN 1334529-08-6

IUPAC Name: 2-(2,4,6-trimethylphenyl)-2H,5H,6H,7H-4λ⁵-pyrrolo[2,1-c][1,2,4]triazol-4-ylium perchlorate
Synonyms: 2-(2,4,6-trimethylphenyl)-5H,6H,7H-4λ⁵-pyrrolo[2,1-c][1,2,4]triazol-4-ylium perchlorate
Molecular Weight (g/mol): 327.77
Molecular Formula: C14H18ClN3O4
InChi Key: LWDOZOIYYGPFSI-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CC1=CC(C)=C(N2C=[N+]3CCCC3=N2)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results