missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium Perchlorate 98.0+%, TCI America™

Catalog No. D3982100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3982100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3982100MG Supplier TCI America Supplier No. D3982100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1334529-08-6
Molecular Formula C14H18ClN3O4
Molecular Weight (g/mol) 327.765
InChI Key LWDOZOIYYGPFSI-UHFFFAOYSA-M
Synonym 6,7-Dihydro-2-mesityl-5H-pyrrolo[2,1-c]-1,2,4-triazolium Perchlorate
PubChem CID 49758092
IUPAC Name 2-(2,4,6-trimethylphenyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-4-ium;perchlorate
SMILES CC1=CC(=C(C(=C1)C)N2C=[N+]3CCCC3=N2)C.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 202°C
Color White
Quantity 100 mg
UN Number 1479
Formula Weight 327.77
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dihydro-2-(2,4,6-trimethylphenyl)-5H-pyrrolo[2,1-c]-1,2,4-triazolium Perchlorate