accessibility menu, dialog, popup

UserName

CAS RN 1334510-66-5

IUPAC Name: 1H,2H-benzo[d]aceanthrylene-1,2-dione
Synonyms: benzo[d]aceanthrylene-1,2-dione
Molecular Weight (g/mol): 282.3
Molecular Formula: C20H10O2
InChi Key: AYSSMZWWOZPRMT-UHFFFAOYSA-N
SMILES: O=C1C(=O)C2=C3C=CC=CC3=CC3=CC4=CC=CC=C4C1=C23
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results