missing translation for 'onlineSavingsMsg'
Learn More

Cyclopenta[fg]tetracene-1,2-dione, TCI America™

Catalog No. C2669200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2669200MG 200 mg
C26691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2669200MG Supplier TCI America Supplier No. C2669200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1334510-66-5
Molecular Formula C20H10O2
Molecular Weight (g/mol) 282.298
InChI Key AYSSMZWWOZPRMT-UHFFFAOYSA-N
Synonym Cyclopenta[fg]naphthacene-1,2-dione
PubChem CID 91972158
SMILES C1=CC=C2C(=C1)C=C3C=C4C=CC=CC4=C5C3=C2C(=O)C5=O

Specifications

Color Blue-Purple
Quantity 200 mg
Formula Weight 282.30
Physical Form Crystalline Powder
Chemical Name or Material Cyclopenta[fg]tetracene-1,2-dione