accessibility menu, dialog, popup

UserName

H3CNCH3OH

CAS RN 13330-96-6

IUPAC Name: 4-(dimethylamino)butan-1-ol
Synonyms: 4-(dimethylamino)butan-1-ol
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: QCTOLMMTYSGTDA-UHFFFAOYSA-N
SMILES: CN(C)CCCCO
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results